2-Iodo-m-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002976-25G 002976-5G
FLUO002976-25GEA 0 GBP
FLUO002976-25G FLUO002976-5G
2-Iodo-m-xylene
2-Iodo-m-xylene
Formula: C₈H₉I
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 11,2 °C
Density: 1,6518 g/cm³ (20 °C)
MDL Number: MFCD00019012
CAS Number: 608-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR