3-Fluoro-5-(trifluoromethyl)benzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006413-5G 006413-25G 006413-1G
FLUO006413-5GEA 0 GBP
FLUO006413-5G FLUO006413-25G FLUO006413-1G
3-Fluoro-5-(trifluoromethyl)benzyl alcohol
3-Fluoro-5-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆F₄O
MW: 194.13 g/mol
MDL Number: MFCD00061169
CAS Number: 184970-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR