4-(Trifluoromethyl)benzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(4-(Trifluoromethyl)phenyl)methanol, 4-(Hydroxymethyl)benzotrifluoride
002182-100G
002182-25G
002182-5G
FLUO002182-100GEA
0
GBP
FLUO002182-100G
FLUO002182-25G
FLUO002182-5G
4-(Trifluoromethyl)benzyl alcohol
4-(Trifluoromethyl)benzyl alcohol
Formula:
C₈H₇F₃O MW: 176.14 g/mol Boiling Pt: 214 °C (1013 hPa) Melting Pt: 23 °C Density: 1,28 g/cm³ (20 °C) |
MDL Number:
MFCD00004661 CAS Number: 349-95-1 EINECS: 206-494-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...