2-(Trifluoromethyl)benzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001975-10G
001975-100G
001975-25G
FLUO001975-10GEA
0
GBP
FLUO001975-10G
FLUO001975-100G
FLUO001975-25G
2-(Trifluoromethyl)benzyl alcohol
2-(Trifluoromethyl)benzyl alcohol
Formula:
C₈H₇F₃O Boiling Pt: 191 °C (1013 hPa) Melting Pt: 4 °C Density: 1,32 g/cm³ (20 °C) |
MDL Number:
MFCD00004621 CAS Number: 346-06-5 EINECS: 206-467-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...