3,5-Bis(trifluoromethyl)anisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Methoxy-3,5-bis(trifluoromethyl)benzene

005567-5G 005567-1G
FLUO005567-5GEA 0 GBP
FLUO005567-5G FLUO005567-1G
3,5-Bis(trifluoromethyl)anisole
3,5-Bis(trifluoromethyl)anisole
Formula: C₉H₆F₆O
Boiling Pt: 158…160 °C (1013 hPa)
Density: 1,4192 g/cm³ (20 °C)
MDL Number: MFCD00728736
CAS Number: 349-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR