α,α,α,α',α',α'-Hexafluoro-3,5-xylidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

004997-100G 004997-500G
FLUO004997-100GEA 0 GBP
FLUO004997-100G FLUO004997-500G
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine
Formula: C₈H₅F₆N
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.4753 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00000394
CAS Number: 328-74-5
EINECS: 206-335-0
UN: 2810
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR