α,α,α,α',α',α'-Hexafluoro-2,5-xylidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,5-Bis(trifluoromethyl)aniline

004996-25G 004996-5G
FLUO004996-25GEA 0 GBP
FLUO004996-25G FLUO004996-5G
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine
Formula: C₈H₅F₆N
MW: 229.12 g/mol
Boiling Pt: 173…177 °C (1013 hPa)
Density: 1.479 g/cm³ (20 °C)
MDL Number: MFCD00074940
CAS Number: 328-93-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR