3,5-Bis(trifluoromethyl)benzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002098-100G 002098-500G 002098-25G
FLUO002098-100GEA 0 GBP
FLUO002098-100G FLUO002098-500G FLUO002098-25G
3,5-Bis(trifluoromethyl)benzoic acid
3,5-Bis(trifluoromethyl)benzoic acid
Formula: C₉H₄F₆O₂
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 140…144 °C
Density: 1,42 g/cm³ (20 °C)
MDL Number: MFCD00000388
CAS Number: 725-89-3
EINECS: 211-970-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR