2,5-Bis(trifluoromethyl)benzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010242-1G 010242-5G
FLUO010242-1GEA 0 GBP
FLUO010242-1G FLUO010242-5G
2,5-Bis(trifluoromethyl)benzoic acid
2,5-Bis(trifluoromethyl)benzoic acid
Formula: C₉H₄F₆O₂
MW: 258.12 g/mol
MDL Number: MFCD00013249
CAS Number: 42580-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR