2,4-Bis(trifluoromethyl)benzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001906-1G 001906-5G
FLUO001906-1GEA 0 GBP
FLUO001906-1G FLUO001906-5G
2,4-Bis(trifluoromethyl)benzoic acid
2,4-Bis(trifluoromethyl)benzoic acid
Formula: C₉H₄F₆O₂
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 111…112 °C
MDL Number: MFCD00042496
CAS Number: 32890-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR