3,5-Bis(trifluoromethyl)benzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002100-25G 002100-100G 002100-10G
FLUO002100-25GEA 0 GBP
FLUO002100-25G FLUO002100-100G FLUO002100-10G
3,5-Bis(trifluoromethyl)benzoyl chloride
3,5-Bis(trifluoromethyl)benzoyl chloride
Formula: C₉H₃ClF₆O
Boiling Pt: 80…81 °C (24 mmHg)
Density: 1.53 g/cm³ (19 °C)
MDL Number: MFCD00000387
CAS Number: 785-56-8
EINECS: 212-322-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR