3,5-Bis(trifluoromethyl)benzoyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
3,5-Bis(trifluoromethyl)benzene-1-carbonyl chloride, 3,5-bis(trifluoromethyl)benzoylchloride
A11401.06
A11401.14
A11401.06EA
34.2
GBP
A11401.06
A11401.14
3,5-Bis(trifluoromethyl)benzoyl chloride ≥97%
3,5-Bis(trifluoromethyl)benzoyl chloride
Purity: 97%
CAS No.: 785-56-8
Molecular Formula: C9H3ClF6O
Formula Weight: 276.57
Flash Point: 72°(161°F)
Refractive Index: 1.4332
Boiling Point: 65-67°/12mm
Beilstein Reference: 2593440
UN No.: 3265
MDL No.: MFCD00000387
Formula:
C₉H₃ClF₆O Boiling Pt: 65…67 °C (12 mmHg) Density: 1.526 Flash Pt: 72 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000387 CAS Number: 785-56-8 EINECS: 212-322-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...