2,4-Bis(trifluoromethyl)benzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006017-1G 006017-5G 006017.10G
FLUO006017-1GEA 0 GBP
FLUO006017-1G FLUO006017-5G FLUO006017.10G
2,4-Bis(trifluoromethyl)benzoyl chloride
2,4-Bis(trifluoromethyl)benzoyl chloride
Formula: C₉H₃ClF₆O
MDL Number: MFCD00061227
CAS Number: 53130-43-1
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR