4'-(Trifluoromethyl)acetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002180-25G
002180-100G
FLUO002180-25GEA
0
GBP
FLUO002180-25G
FLUO002180-100G
4'-(Trifluoromethyl)acetophenone
4'-(Trifluoromethyl)acetophenone
Formula:
C₉H₇F₃O Boiling Pt: 205 °C (1013 hPa) Melting Pt: 26…29 °C Density: 0,924 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000401 CAS Number: 709-63-7 EINECS: 211-913-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...