3'-(Trifluoromethyl)acetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: m-(Trifluoromethyl)acetophenone, 1-[3-(Trifluoromethyl)phenyl]ethan-1-one

001546-25G 001546-100G
FLUO001546-25GEA 0 GBP
FLUO001546-25G FLUO001546-100G
3'-(Trifluoromethyl)acetophenone
3'-(Trifluoromethyl)acetophenone
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 198
Density: 1.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000391
CAS Number: 349-76-8
EINECS: 206-490-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR