3'-(Trifluoromethyl)acetophenone ≥98%
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Purity: 98+%
CAS No.: 349-76-8
Molecular Formula: C9H7F3O
Formula Weight: 188.15
Flash Point: 83°(181°F)
Refractive Index: 1.4635
Odour: Characteristic
Boiling Point: 198-200°
Beilstein Reference: 640151
MDL No.: MFCD00000391
Formula:
C₉H₇F₃O Boiling Pt: 198…200 °C Density: 1.260 Flash Pt: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000391 CAS Number: 349-76-8 EINECS: 206-490-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...