2'-(Trifluoromethyl)acetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001970-5G
001970-25G
FLUO001970-5GEA
0
GBP
FLUO001970-5G
FLUO001970-25G
2'-(Trifluoromethyl)acetophenone
2'-(Trifluoromethyl)acetophenone
Formula:
C₉H₇F₃O Boiling Pt: 163 °C (1013 hPa) Melting Pt: 15…17 °C Density: 1,25 g/cm³ (20 °C) |
MDL Number:
MFCD00000378 CAS Number: 17408-14-9 EINECS: 241-434-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...