3'-(Trifluoromethyl)propiophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(3-(Trifluoromethyl)phenyl)propan-1-one, m-(Trifluoromethyl)propiophenone

007659-10G 007659-25G
FLUO007659-10GEA 0 GBP
FLUO007659-10G FLUO007659-25G
3'-(Trifluoromethyl)propiophenone
3'-(Trifluoromethyl)propiophenone
Formula: C₁₀H₉F₃O
MDL Number: MFCD00039227
CAS Number: 1533-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR