4-Fluoro-o-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

002578-5G 002578-25G
FLUO002578-5GEA 0 GBP
FLUO002578-5G FLUO002578-25G
4-Fluoro-o-xylene
4-Fluoro-o-xylene
Formula: C₈H₉F
Boiling Pt: 143…144 °C (730 mmHg)
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00039217
CAS Number: 452-64-2
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR