2-Bromo-4,6-xylidine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 6-Bromo-2,4-dimethylaniline, 6-Bromo-2,4-xylidine, 2-Bromo-4,6-dimethylaniline

M-Clarity™ quality level = MQ100

528862-10G 528862-1G
SIAL528862-10GEA 120 GBP
SIAL528862-10G SIAL528862-1G
2-Bromo-4,6-xylidine, Sigma-Aldrich®
2-Bromo-4,6-xylidine
Formula: C₈H₁₀BrN
MW: 200.08 g/mol
MDL Number: MFCD00047826
CAS Number: 41825-73-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR