5-Bromo-2-fluoro-m-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

009060-5G 009060-25G
FLUO009060-5GEA 0 GBP
FLUO009060-5G FLUO009060-25G
5-Bromo-2-fluoro-m-xylene
5-Bromo-2-fluoro-m-xylene
Formula: C₈H₈BrF
MDL Number: MFCD01320701
CAS Number: 99725-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR