1-Bromo-3,5-bis(trifluoromethyl)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3,5-Bis(trifluoromethyl)-1-bromobenzene, 1-Bromo-3,5-bis(trifluoromethyl)benzene, 5-Bromo-α,α,α,β,β,β-Hexafluoro-m-xylene, 1,3-Bis(trifluoromethyl)-5-bromobenzene, 3,5-Bis(trifluoromethyl)bromobenzene, 5-Bromo-α,α,α,α',α',α'-Hexafluoro-m-xylene

002104-100G 002104-500G 002104-25G
FLUO002104-100GEA 0 GBP
FLUO002104-100G FLUO002104-500G FLUO002104-25G
1-Bromo-3,5-bis(trifluoromethyl)benzene
1-Bromo-3,5-bis(trifluoromethyl)benzene
Formula: C₈H₃BrF₆
Boiling Pt: 152…155 °C (1013 hPa)
Melting Pt: –16 °C
Density: 1,7048 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000381
CAS Number: 328-70-1
EINECS: 206-334-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR