3-Bromo-o-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

018326-100G 018326-25G
FLUO018326-100GEA 0 GBP
FLUO018326-100G FLUO018326-25G
3-Bromo-o-xylene
3-Bromo-o-xylene
Formula: C₈H₉Br
Boiling Pt: 214 °C (1013 hPa)
Density: 1.365 g/cm³ (20 °C)
MDL Number: MFCD00000069
CAS Number: 576-23-8
EINECS: 209-398-2
UN: 1701
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR