4,5-Dibromo-o-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

013144-1G 013144-5G
FLUO013144-1GEA 0 GBP
FLUO013144-1G FLUO013144-5G
4,5-Dibromo-o-xylene
4,5-Dibromo-o-xylene
Formula: C₈H₈Br₂
MDL Number: MFCD00082743
CAS Number: 24932-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR