Thymol blue, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

114545-25G 114545-5G
SIAL114545-25GEA 129 GBP
SIAL114545-25G SIAL114545-5G
Thymol blue, Sigma-Aldrich®
Thymol blue
Formula: C₂₇H₃₀O₅S
Melting Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD00005869
CAS Number: 76-61-9
EINECS: 200-973-3
Merck Index: 12,09541

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR