Dibenzylamine 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

112612500 112610010
ACRO112612500EA 91.6 GBP
ACRO112612500 ACRO112610010
Dibenzylamine 98%
Dibenzylamine
Formula: C₆H₅CH₂NHCH₂C₆H₅
Boiling Pt: 300 °C (760 mmHg)
Melting Pt: –26 °C
Density: 1.02 g/cm³
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00004770
CAS Number: 103-49-1
EINECS: 203-117-7
UN: 2735
ADR: 8,III
Merck Index: 13,03035
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c17e97ec30680cc9e622a04bff55a48e1b301831&vid=c74bd48fc42a0747a1603d963976594b20a27385&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5740 to 1.5760 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR