2,3,4,5,6-Pentafluorobenzophenone ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

A13713.06 A13713.14
A13713.06EA 117 GBP
A13713.06 A13713.14
2,3,4,5,6-Pentafluorobenzophenone ≥98%
2,3,4,5,6-Pentafluorobenzophenone

Purity: 98% CAS No.: 1536-23-8 Molecular Formula: C13H5F5O Formula Weight: 272.18 Flash Point: 110°(230°F) Melting Point: 36-39° Boiling Point: 93°/0.2mm Beilstein Reference: 2059098 MDL No.: MFCD00000294

Formula: C₁₃H₅F₅O
MW: 272.17 g/mol
Boiling Pt: 93 °C (0.2 mmHg)
Melting Pt: 36…39 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000294
CAS Number: 1536-23-8
EINECS: 216-258-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR