4-Fluorobenzophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
003845-100G
003845-25G
FLUO003845-100GEA
0
GBP
FLUO003845-100G
FLUO003845-25G
4-Fluorobenzophenone
4-Fluorobenzophenone
Formula:
C₁₃H₉FO MW: 200.21 g/mol Boiling Pt: 302 °C (1013 hPa) Melting Pt: 49 °C |
MDL Number:
MFCD00000352 CAS Number: 345-83-5 EINECS: 206-463-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...