2,5-Difluorobenzophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001936-5G
001936-25G
FLUO001936-5GEA
0
GBP
FLUO001936-5G
FLUO001936-25G
2,5-Difluorobenzophenone
2,5-Difluorobenzophenone
Formula:
C₁₃H₈F₂O MW: 218.2 g/mol |
MDL Number:
MFCD00009899 CAS Number: 85068-36-6 EINECS: 285-298-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...