2,4-Difluorobenzophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001919-5G
001919-25G
FLUO001919-5GEA
0
GBP
FLUO001919-5G
FLUO001919-25G
2,4-Difluorobenzophenone
2,4-Difluorobenzophenone
Formula:
C₁₃H₈F₂O MW: 218.2 g/mol Boiling Pt: 120 °C (2 mmHg) Melting Pt: 33 °C Density: 1,26 g/cm³ (20 °C) |
MDL Number:
MFCD00000331 CAS Number: 85068-35-5 EINECS: 285-297-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...