2,6-Difluorobenzophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001959-5G 001959-25G
FLUO001959-5GEA 0 GBP
FLUO001959-5G FLUO001959-25G
2,6-Difluorobenzophenone
2,6-Difluorobenzophenone
Formula: C₁₃H₈F₂O
MW: 218.2 g/mol
Boiling Pt: 120 °C (2 mmHg)
Density: 1,24 g/cm³ (20 °C)
MDL Number: MFCD00009895
CAS Number: 59189-51-4
EINECS: 261-650-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR