2,2-Diphenylpropiononitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,2-Diphenylpropionitrile

001097-25G 001097-1KG 001097-100G
FLUO001097-25GEA 0 GBP
FLUO001097-25G FLUO001097-1KG FLUO001097-100G
2,2-Diphenylpropiononitrile
2,2-Diphenylpropiononitrile
Formula: (C₆H₅)₂C(CN)CH₃
Boiling Pt: 145 °C (1013 hPa)
Density: 1.0671 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001846
CAS Number: 5558-67-8
EINECS: 226-925-1
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e27ce7e18aa15b0826d9325a7b9518b82352b767&vid=e27ce7e18aa15b0826d9325a7b9518b82352b767&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR