Bromocresol green, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
17470-25G
17470-5G
SIAL17470-25GEA
248
GBP
SIAL17470-25G
SIAL17470-5G
Bromocresol green, Sigma-Aldrich®
Bromocresol green
Formula:
C₂₁H₁₄Br₄O₅S Melting Pt: 217…218 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005874 CAS Number: 76-60-8 EINECS: 200-972-8 Merck Index: 13,01370 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...