Pentylbenzene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
|
Synonyms:
1-Phenylpentane, Amylbenzene
M-Clarity™ quality level = MQ200
113174-100G
113174-25G
SIAL113174-100GEA
272
GBP
SIAL113174-100G
SIAL113174-25G
Pentylbenzene, Sigma-Aldrich®
Pentylbenzene
Formula:
C₁₁H₁₆ MW: 148.25 g/mol Boiling Pt: 205 °C (1013 hPa) Melting Pt: –1 °C Density: 0.8594 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009502 CAS Number: 538-68-1 EINECS: 208-701-5 UN: 3082 ADR: 9,III Merck Index: 13,00604 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...