Toluene

Supplier: BARRETTINE INDUSTRIAL
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

TOLU005 TOLU205 TOLU025
BARRTOLU005EA 0 GBP
BARRTOLU005 BARRTOLU205 BARRTOLU025
Toluene
Toluene
Formula: C₆H₅CH₃
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR