o-Toluenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Toluene-o-sulphonyl chloride, Toluene-2-sulphonyl chloride, 2-Toluenesulphonyl chloride

013862-25G FL13862.100 013862-5G
FLUO013862-25GEA 0 GBP
FLUO013862-25G CHMPFL13862.100 FLUO013862-5G
o-Toluenesulphonyl chloride
o-Toluenesulphonyl chloride
Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 10.2 °C
Density: 1.3383 g/cm³ (20 °C)
MDL Number: MFCD00024874
CAS Number: 133-59-5
EINECS: 205-113-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR