4-Toluenethiol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Mercapto-4-methylbenzene, 4-Methylbenzene-1-thiol, 4-Thiocresol, 4-Methylbenzenethiol, p-Toluenethiol, p-Thiocresol

019192-50G 019192-500G
FLUO019192-50GEA 0 GBP
FLUO019192-50G FLUO019192-500G
4-Toluenethiol
4-Toluenethiol
Formula: C₇H₈S
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: 39…42 °C
Density: 1.022 g/cm³ (51 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00004851
CAS Number: 106-45-6
EINECS: 203-399-1
UN: 2811
ADR: 6.1,III
Merck Index: 12,09461

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR