4-Ethylthiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003726.1G 003726-5G 003726-25G
FLUO003726.1GEA 0 GBP
FLUO003726.1G FLUO003726-5G FLUO003726-25G
4-Ethylthiophenol
4-Ethylthiophenol
Formula: C₈H₁₀S
Boiling Pt: 119…120 °C (30 mmHg)
MDL Number: MFCD00040229
CAS Number: 4946-13-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR