3,5-Dihydroxytoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005921-25G 005921-5G 005921-100G
FLUO005921-25GEA 0 GBP
FLUO005921-25G FLUO005921-5G FLUO005921-100G
3,5-Dihydroxytoluene
3,5-Dihydroxytoluene
Formula: C₇H₈O₂
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 109 °C
Density: 1.2895 g/cm³ (20 °C)
Flash Pt: 159 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002291
CAS Number: 504-15-4
EINECS: 207-984-2
Merck Index: 12,06995

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR