m-Cresol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1-Hydroxy-3-methylbenzene, 3-Methylphenol, 3-Hydroxytoluol, 3-Hydroxytoluene

M-Clarity™ quality level = MQ100

W353000-10KG-K W353000-1KG-K W353000-25KG-K
SIALW353000-10KG-KEA 492 GBP
SIALW353000-10KG-K SIALW353000-1KG-K SIALW353000-25KG-K
m-Cresol, Sigma-Aldrich®
m-Cresol
Formula: CH₃C₆H₄OH
MW: 108.14 g/mol
Boiling Pt: 202.2
Melting Pt: 11.5
Density: 1.0336 g/cm³ (20 °C)
Flash Pt: 73
Storage Temperature: Ambient
MDL Number: MFCD00002302
CAS Number: 108-39-4
EINECS: 203-577-9
UN: 2076
ADR: 6.1 II
Merck Index: 13,02604
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce465c84cc8b7c230526b22e6f830300b605d109&vid=72c08ae3da371936f9349196418a240331cb79ce&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR