4-Methoxybenzyl alcohol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Anisyl alcohol, p-Anisyl Alcohol, 4-(Hydroxymethyl)anisole, Anise alcohol, (4-Methoxyphenyl)methanol

M-Clarity™ quality level = MQ100

W209910-100G-K W209910-1KG-K
SIALW209910-100G-KEA 198 GBP
SIALW209910-100G-K SIALW209910-1KG-K
4-Methoxybenzyl alcohol, Sigma-Aldrich®
4-Methoxybenzyl alcohol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: 20…24 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004653
CAS Number: 105-13-5
EINECS: 203-273-6
Merck Index: 13,00608

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR