3-Nitrobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Nitrophenyl)methanol

008826-10G 008826-50G
FLUO008826-10GEA 0 GBP
FLUO008826-10G FLUO008826-50G
3-Nitrobenzyl alcohol
3-Nitrobenzyl alcohol
Formula: C₇H₇NO₃
Boiling Pt: 317 °C (1013 hPa)
Melting Pt: 26…27 °C
Density: 1.3 g/cm³ (20 °C)
MDL Number: MFCD00007273
CAS Number: 619-25-0
EINECS: 210-588-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR