N-Methyl-O-toluidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

009654-10G 009654-25G
FLUO009654-10GEA 0 GBP
FLUO009654-10G FLUO009654-25G
N-Methyl-O-toluidine
N-Methyl-o-toluidine
Formula: C₈H₁₁N
Boiling Pt: 206…208 °C (1013 hPa)
Density: 0.973 g/cm³ (15 °C)
MDL Number: MFCD00025627
CAS Number: 611-21-2
EINECS: 210-260-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR