3-Methoxybenzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

008880-5G 008880-25G
FLUO008880-5GEA 0 GBP
FLUO008880-5G FLUO008880-25G
3-Methoxybenzylamine
3-Methoxybenzylamine
Formula: C₈H₁₁NO
Boiling Pt: 238 °C (1013 hPa)
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00008115
CAS Number: 5071-96-5
EINECS: 225-779-6
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR