2-Methoxybenzylamine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ100
159883-25G
159883-5G
SIAL159883-25GEA
139
GBP
SIAL159883-25G
SIAL159883-5G
2-Methoxybenzylamine, Sigma-Aldrich®
2-Methoxybenzylamine
Formula:
C₈H₁₁NO Boiling Pt: 227 °C (1013 hPa) Density: 1.05 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008110 CAS Number: 6850-57-3 EINECS: 229-941-7 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...