2-Methoxybenzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: (2-Methoxyphenyl)methanamine, 2-(Aminomethyl)anisole, o-Anisylamine

008875-10G 008875-50G
FLUO008875-10GEA 0 GBP
FLUO008875-10G FLUO008875-50G
2-Methoxybenzylamine
2-Methoxybenzylamine
Formula: C₈H₁₁NO
Boiling Pt: 227 °C (1013 hPa)
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008110
CAS Number: 6850-57-3
EINECS: 229-941-7
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR