3-Nitro-o-toluidine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
3-Nitro-o-methylaniline, 3-Nitro-2-methylaniline, 2-Methyl-3-nitroaniline, 2-Amino-6-nitrotoluene
M-Clarity™ quality level = MQ100
115843-100G
115843-25G
SIAL115843-100GEA
145
GBP
SIAL115843-100G
SIAL115843-25G
3-Nitro-o-toluidine, Sigma-Aldrich®
3-Nitro-o-toluidine
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Boiling Pt: 305 Melting Pt: 87…89 °C Density: 0.33 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007731 CAS Number: 603-83-8 EINECS: 210-059-6 UN: 2660 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...