p-Toluidine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
4-Aminotoluene, 4-Methylaniline, 4-Toluidine
M-Clarity™ quality level = MQ200
461121-1KG
461121-250G
SIAL461121-1KGEA
115
GBP
SIAL461121-1KG
SIAL461121-250G
p-Toluidine, Sigma-Aldrich®
p-Toluidine
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 201 Melting Pt: 43 Density: 1.046 g/cm³ (20 °C) Flash Pt: 87 Storage Temperature: Ambient |
MDL Number:
MFCD00007906 CAS Number: 106-49-0 EINECS: 203-403-1 UN: 3451 ADR: 6.1 II Merck Index: 12,09674 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...