Phenethylamine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
PEA, β-Phenylethylamine, 2-Phenylethylamine
M-Clarity™ quality level = MQ200
128945-1L
128945-250ML
SIAL128945-1LEA
159
GBP
SIAL128945-1L
SIAL128945-250ML
Phenethylamine, Sigma-Aldrich®
Phenethylamine
Formula:
C₆H₅CH₂CH₂NH₂ MW: 121.18 g/mol Boiling Pt: 200 Melting Pt: –60 °C Density: 0.9639 g/cm³ (20 °C) Flash Pt: 81 Storage Temperature: Ambient |
MDL Number:
MFCD00008184 CAS Number: 64-04-0 EINECS: 200-574-4 UN: 2735 ADR: 8 I Merck Index: 12,07371 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...