2-Methyl-4-nitrophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009538-1G 009538-5G
FLUO009538-1GEA 0 GBP
FLUO009538-1G FLUO009538-5G
2-Methyl-4-nitrophenyl isothiocyanate
2-Methyl-4-nitrophenyl isothiocyanate
Formula: C₈H₆N₂O₂S
MW: 194.21 g/mol
MDL Number: MFCD00060542
CAS Number: 135805-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR